Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FFAR3/GPR41 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10562R10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10562R10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10562R10 Supplier R&D Systems Supplier No. FAB10562R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FFAR3/GPR41 Monoclonal specifically detects FFAR3/GPR41 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FFAR3/GPR41
Applications Flow Cytometry
Classification Monoclonal
Clone 2590G
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias FFA3, FFA3R, free fatty acid receptor 3, G protein-coupled receptor 41, GPR41, GPR41FFA3R, G-protein coupled receptor 41
Host Species Rabbit
Immunogen Human FFAR3/GPR41 synthetic peptide
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2865
Reconstitution Reconstitution default not known
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.