Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FFAR3/GPR41 MAb (Clone 2590G), R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | FFAR3/GPR41 |
---|---|
Clone | 2590G |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/106 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10562SP
![]() |
R&D Systems
MAB10562SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10562100
![]() |
R&D Systems
MAB10562100 |
100 μg |
Each for $467.50
|
|
|||||
Description
FFAR3/GPR41 Monoclonal antibody specifically detects FFAR3/GPR41 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
FFAR3/GPR41 | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/106 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
2865 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
2590G | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Rabbit | |
Human | |
FFA3, FFA3R, free fatty acid receptor 3, G protein-coupled receptor 41, GPR41, GPR41FFA3R, G-protein coupled receptor 41 | |
Synthetic peptide containing human FFAR3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title