Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3451SP
This item is not returnable.
View return policy
Description
FGF-10 Monoclonal specifically detects FGF-10 in Human samples. It is validated for ELISA.Specifications
FGF-10 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
FGF10, FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, KGF2, KGF-2, produced by fibroblasts of urinary bladder lamina propria | |
E.coli-derived recombinant human FGF-10, Cys37-Ser208, Accession No. O15520 | |
25 μg | |
2255.0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG3 |
ELISA | |
963521 | |
ELISA | |
O15520 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human FGF-10 in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction