Learn More
Human FGF-11 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | FGF-11 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL |
| Gene Alias | FGF11, FGF-11, FHF-3, FHF3Fibroblast growth factor homologous factor 3, fibroblast growth factor 11, FLJ16061, MGC102953, MGC45269 |
| Gene Symbols | FGF11 |
| Host Species | Sheep |
| Immunogen | E. coli-derived recombinant human FGF-11 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.