Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF-12 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $514.50
Specifications
Antigen | FGF-12 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
FGF-12 Polyclonal specifically detects FGF-12 in Human samples. It is validated for Western Blot.Specifications
FGF-12 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
FGF12, FGF-12, FGF12B, FHF1, FHF-1, FHF1Fibroblast growth factor homologous factor 1, fibroblast growth factor 12, fibroblast growth factor 12B, fibroblast growth factor FGF-12b, Myocyte-activating factor | |
FGF12 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P61328 | |
2257 | |
E. coli-derived recombinant human FGF-12 Glu67-Thr243 Accession # P61328 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human FGF-12 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human (rh) FGF-13B is observed, 10% cross-reactivity with rhFGF-11 is observed and less than 1% cross-reactivity with rhFGF acidic, rhFGF basic, rhFGF-3, rhFGF-4, rhFGF-6, rhFGF-7, rhFGF-9, rhFGF-10, rhFGF-16, rhFGF-17, rhFGF-18, rhFGF-19, rhFGF-20, rhFGF-23, recombinant mouse (rm) FGF-15, and rmFGF-8b is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title