Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF-16 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | FGF-16 |
---|---|
Clone | 209511 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
FGF-16 Monoclonal specifically detects FGF-16 in Human samples. It is validated for Western Blot.Specifications
FGF-16 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O43320 | |
8823 | |
E. coli-derived recombinant human FGF-16 Ala2-Arg207 Accession # O43320 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human FGF-16 in direct ELISAs and Western blots. In direct ELISAs, less than 25% cross-reactivity with recombinant human (rh) FGF-3, -5, -8c, and -11 is observed and no cross-reactivity with rhFGF acidic, rhFGF basic, rhFGF-4, -6, -7, -8, -9, -10, -12, -13, -17, -18, or -19 is observed. |
209511 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FGF16, FGF-16, fibroblast growth factor 16 | |
FGF16 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title