Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FGF-18 Protein

Catalog No. 8988F18050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human FGF-18 Protein is derived from E. coli. The Recombinant Human FGF-18 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number O76093
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and Trehalose.
Gene ID (Entrez) 8817
Molecular Weight (g/mol) Observed M.W.: 19-24kDa, reducing conditions, Theoretical M.W.: 21kDa
Quantity 50 μg
Source E. coli-derived human FGF-18 protein Glu28-Ala207,with a N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias FGF18, FGF-18ZFGF5, fibroblast growth factor 18, zFGF5
Common Name FGF18
Gene Symbol FGF18
Biological Activity Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with human FGF RIIIc. The ED50 for this effect is typically 4-24ng/mL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Glu28-Ala207, with a N-terminal Met
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.