Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGF-20 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2547
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2547 100 μg
MAB2547SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2547 Supplier R&D Systems Supplier No. MAB2547
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

FGF-20 Monoclonal specifically detects FGF-20 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen FGF-20
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 272317
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9NP95
Gene Alias FGF20, FGF-20, fibroblast growth factor 20
Gene Symbols FGF20
Host Species Rat
Immunogen E. coli-derived recombinant human FGF-20 Met1-Thr211 Accession # Q9NP95
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26281
Test Specificity Detects human FGF-20 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) FGF-3, -4, -5, -6, -7, -9, -10, -11, -12, -16, -17, -18, -19, -23, recombinant mouse FGF-8b, -8c, -15, -23, rhFGF acidic, or rhFGF basic is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.