Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF-21 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC25372U100UG
This item is not returnable.
View return policy
Description
FGF-21 Monoclonal specifically detects FGF-21 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
FGF-21 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9NSA1 | |
FGF21 | |
E. coli-derived recombinant human FGF-21 His29-Ser209 Accession # Q9NSA1 | |
RUO | |
26291 | |
Human | |
IgG2b |
Flow Cytometry | |
461804 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FGF21, FGF-21, fibroblast growth factor 21 | |
Mouse | |
100 μg | |
Primary | |
Detects human FGF-21 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse FGF-21, recombinant human (rh) FGF-3, -4, -6 or -10 is observed and no cross-reactivity with rhFGF-5 or rhFGF-9 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction