Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGF-21 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB25372
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB25372 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB25372 Supplier R&D Systems Supplier No. MAB25372
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGF-21 Monoclonal specifically detects FGF-21 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen FGF-21
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 461804
Concentration LYOPH
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 15-30 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9NSA1
Gene Alias FGF21, FGF-21, fibroblast growth factor 21
Gene Symbols FGF21
Host Species Mouse
Immunogen E. coli-derived recombinant human FGF-21 His29-Ser209 Accession # Q9NSA1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26291
Test Specificity Detects human FGF-21 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse FGF-21, recombinant human (rh) FGF-3, -4, -6 or -10 is observed and no cross-reactivity with rhFGF-5 or rhFGF-9 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.