Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF-21 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $585.00
Specifications
Antigen | FGF-21 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, ELISA |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Description
FGF-21 Polyclonal specifically detects FGF-21 in Human samples. It is validated for Western Blot, ELISA.Specifications
FGF-21 | |
Western Blot 0.1 ug/mL, ELISA | |
Polyclonal | |
Goat | |
Human | |
FGF21, FGF-21, fibroblast growth factor 21 | |
FGF21 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Q9NSA1 | |
26291 | |
E. coli-derived recombinant human FGF-21 His29-Ser209 Accession # Q9NSA1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human FGF-21 in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant mouse (rm) FGF-21 is observed and less than 2% cross-reactivity with recombinant human (rh) FGF-3, rhFGF-4, rhFGF-5, rhFGF-6, rhFGF-7, rmFGF-8b, rhFGF-9, rhFGF-10, rhFGF-11, rhFGF-12, rmFGF-15, rhFGF-16, rhFGF-17, rhFGF-18, rhFGF-19, rhFGF-20, rhFGF-23, rhFGF acidic, and rhFGF basic is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title