Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGF-21 Antibody, R&D Systems™
GREENER_CHOICE

Goat Monoclonal Antibody

$214.00 - $549.50

Specifications

Antigen FGF-21
Clone 40020A
Dilution Neutralization 0.5-3 ug/mL
Applications Neutralization
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB25373SP
GREENER_CHOICE
View Documents
R&D Systems
MAB25373SP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB25373100
GREENER_CHOICE
View Documents
R&D Systems
MAB25373100
100 μg
Each for $549.50
Only null left
Add to Cart
 
Description

Description

FGF-21 Monoclonal specifically detects FGF-21 in Human samples. It is validated for Neutralization.
Specifications

Specifications

FGF-21
Neutralization 0.5-3 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
FGF21, FGF-21, fibroblast growth factor 21
fgf21
IgG2b
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
40020A
Neutralization
Unconjugated
Goat
Human
Q9NSA1
26291
E. coli-derived recombinant human FGF-21 His29-Ser209 Accession # Q9NSA1
Primary
Reconstitue at 0.5mg/mL in sterile PBS.
Detects human FGF-21 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.