Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF-22 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | FGF-22 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
FGF-22 Polyclonal specifically detects FGF-22 in Human samples. It is validated for Western Blot.Specifications
FGF-22 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
FGF22, FGF-22, fibroblast growth factor 22 | |
FGF22 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9HCT0 | |
27006 | |
E. coli-derived recombinant human FGF-22 Thr23-Ser170 Accession # Q9HCT0 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human FGF-22 in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) FGF-2, -3, -4, -5, -6, -7, -8b, -9, -10, -11, -12, -13, -16, -17, -18, -19, -20, -23, and recombinant mouse (rm) FGF-8, rmFGF-15, and rhFGF-acidic is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title