Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | FGF-4 |
---|---|
Clone | 26019 |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
FGF-4 Monoclonal specifically detects FGF-4 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
FGF-4 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FGF4, fibroblast growth factor 4, HBGF-4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST | |
FGF4 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
26019 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P08620 | |
2249 | |
E. coli-derived recombinant human FGF-4 Accession # P08620 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
When used in combination with the biotinylated anti-FGF-4 detection antibody (Catalog # BAF235) in sandwich ELISAs, no significant cross-reactivity or interference was observed with recombinant human (rh) FGF acidic, bFGF acidic, rhFGF basic, bFGF basic, rhFGF-5, rhFGF-6, or rhFGF-7 (KGF). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title