Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGF-5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2371SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2371SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2371SP Supplier R&D Systems Supplier No. MAB2371SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGF-5 Monoclonal specifically detects FGF-5 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen FGF-5
Applications Immunohistochemistry
Classification Monoclonal
Clone 221903
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8NF90
Gene Alias FGF5, FGF-5, fibroblast growth factor 5, HBGF-5, Heparin-binding growth factor 5, Smag-82
Gene Symbols FGF5
Host Species Mouse
Immunogen E. coli-derived recombinant human FGF-5 long isoform Glu23-Gly268 (Lys238Asn, Pro245Ser) Accession # Q8NF90
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2250
Test Specificity Detects human FGF-5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) FGF acidic, rhFGF basic, rhFGF-3, -4, -6, -7, -9, -10, -11, -12, -13, -16, -17, -18, -19, -20, -21, -23, recombinant mouse FGF-8b, -8c, -15, or -23 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.