Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
$200.50 - $2020.00
Specifications
Antigen | FGF-9 |
---|---|
Clone | 36912 |
Dilution | Neutralization 0.1-0.6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Neutralization, ELISA |
Classification | Monoclonal |
Description
FGF-9 Monoclonal specifically detects FGF-9 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
FGF-9 | |
Neutralization 0.1-0.6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P31371 | |
2254 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF-9 Met1-Ser208 Accession # P31371 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human FGF-9 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) FGF-4, 5, 6, 7, 16, rhFGF acidic, or rhFGF basic is observed. |
36912 | |
Neutralization, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FGF9, FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3 | |
FGF9 | |
IgG2a | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title