Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF acidic/FGF1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | FGF acidic/FGF1 |
---|---|
Clone | 939902 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
FGF acidic/FGF1 Monoclonal specifically detects FGF acidic/FGF1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FGF acidic/FGF1 | |
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AFGF, alpha, alpha-ECGF, beta-ECGF, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGF acidic, FGF-1, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 | |
FGF1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
939902 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P05230 | |
2246 | |
E. coli-derived recombinant human FGF acidic/FGF1, Phe16-Asp155, Accession # P05230 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human FGF acidic/FGF1 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title