Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGF acidic/FGF1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF232
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF232 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF232 Supplier R&D Systems Supplier No. AF232
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 6 publications

FGF acidic/FGF1 Polyclonal specifically detects FGF acidic/FGF1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Neutralization.

Specifications

Antigen FGF acidic/FGF1
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 μg/mL, Simple Western 2.5 μg/mL, Immunohistochemistry 5-15 μg/mL, Neutralization
Gene Accession No. P05230
Gene Alias AFGF, alpha, alpha-ECGF, beta-ECGF, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGF acidic, FGF-1, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1
Gene Symbols FGF1
Host Species Goat
Immunogen E. coli-derived recombinant human FGF acidic/FGF1, Phe16-Asp155, Accession # P05230
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2246
Test Specificity Detects human FGF acidic/FGF1 in direct ELISAs and Western blots. In direct ELISAs, approximately 100% cross-reactivity with recombinant mouse FGF acidic/FGF1 is observed. Neutralizes the biological activity of rhFGF acidic/FGF1. It will also neutralize the biological activity of rh beta-ECGF and bovine FGF acidic/FGF1, although 2-3 times the amount of IgG is required to neutralize bovine FGF acidic/FGF1 biological activity.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.