Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF basic/FGF2/bFGF Antibody, Novus Biologicals™

Rat Monoclonal Antibody
$177.10 - $485.00
Specifications
Antigen | FGF basic/FGF2/bFGF |
---|---|
Clone | 954824 |
Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2332100
![]() |
R&D Systems
MAB2332100 |
100 μg |
Each for $485.00
|
|
|||||
MAB2332SP
![]() |
R&D Systems
MAB2332SP |
25 μg |
Each for $177.10
|
|
|||||
Description
FGF basic/FGF2/bFGF Monoclonal antibody specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
FGF basic/FGF2/bFGF | |
Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin | |
E. coli-derived human FGF basic/FGF2/bFGF, Pro143-Ser288, Accession # P09038 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
954824 | |
Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rat | |
Human | |
P09038 | |
2247 | |
IgG2a | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title