Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF basic/FGF2/bFGF Antibody, R&D Systems™

Mouse Monoclonal Antibody
$177.10 - $474.50
Specifications
Antigen | FGF basic/FGF2/bFGF |
---|---|
Clone | 10044 |
Dilution | Immunocytochemistry 5-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2333100
![]() |
R&D Systems
MAB2333100 |
100 μg |
Each for $474.50
|
|
|||||
MAB2333SP
![]() |
R&D Systems
MAB2333SP |
25 μg |
Each for $177.10
|
|
|||||
Description
FGF basic/FGF2/bFGF Monoclonal antibody specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for ImmunocytochemistrySpecifications
FGF basic/FGF2/bFGF | |
Immunocytochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin | |
E. coli -derived human FGF basic/FGF2/bFGF, Accession # P09038 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
10044 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P09038 | |
2247 | |
IgG2a | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title