Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGF-BP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1593SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1593SP 25 μg
AF1593 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1593SP Supplier R&D Systems Supplier No. AF1593SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FGF-BP Polyclonal specifically detects FGF-BP in Human samples. It is validated for Western Blot.

Specifications

Antigen FGF-BP
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q14512
Gene Alias 17 kDa heparin-binding growth factor-binding protein, FGFBP, FGF-BP, FGFBP1, FGF-BP1, FGFBP-1, FGFBPFGF-binding protein 1, fibroblast growth factor binding protein 1, fibroblast growth factor-binding protein 1, HBp17, HBP1717 kDa HBGF-binding protein, heparin-binding growth factor binding protein
Gene Symbols FGFBP1
Host Species Goat
Immunogen E. coli-derived recombinant human FGF-BP Lys34-Cys234 Accession # Q14512
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9982
Test Specificity Detects human FGF-BP in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant rat FGF-BP is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.