Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGFR1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $2070.00

Specifications

Antigen FGFR1
Clone 133105
Dilution Western Blot 1 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB658SP
GREENER_CHOICE
View Documents
R&D Systems
MAB658SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB658100
GREENER_CHOICE
View Documents
R&D Systems
MAB658100
100 μg
Each for $561.50
Only null left
Add to Cart
 
MAB658500
GREENER_CHOICE
View Documents
R&D Systems
MAB658500
500 μg
Each for $2,070.00
Only null left
Add to Cart
 
Description

Description

FGFR1 Monoclonal specifically detects FGFR1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FGFR1
Western Blot 1 ug/mL
Monoclonal
Mouse
Human
basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGF R1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, Flt-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2
FGFR1
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
133105
Western Blot
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
2260
Mouse myeloma cell line NS0-derived recombinant human FGF R1 alpha (IIIc)
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human FGF R1 in direct ELISAs and Western blots. Recognizes all isoforms of recombinant human (rh) FGF R1 and shows 10-25% cross-reactivity with various isoforms of rhFGF R2 and rhFGF R3.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.