Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGFR2 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $526.00
Specifications
| Antigen | FGFR2 alpha |
|---|---|
| Clone | 1057909 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11119SP
![]() |
R&D Systems
MAB11119SP |
25 μg |
Each for $214.00
|
|
|||||
MAB11119100
![]() |
R&D Systems
MAB11119100 |
100 μg |
Each for $526.00
|
|
|||||
Description
FGFR2 alpha Monoclonal antibody specifically detects FGFR2 alpha in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
| FGFR2 alpha | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P21802.1 | |
| Chinese Hamster Ovary cell line, CHO-derived human FGFR2 alpha protein, Arg22-Glu377, Accession # P21802.1 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 1057909 | |
| Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 2263 | |
| IgG2b | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title