Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGFR2 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB684A
This item is not returnable.
View return policy
Description
FGFR2 Monoclonal specifically detects FGFR2 in Human samples. It is validated for Flow Cytometry.Specifications
FGFR2 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
FGFR2 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms and Mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms | |
100 Tests | |
Primary | |
Detects alpha isoforms (3 Ig-like domains) and beta isoforms (2 Ig-like domains) of human FGF R2 in direct ELISAs and Western blots. In direct ELISAs, this antibody reacts with all isoforms of rhFGF R2 but shows approximately a 4-fold preference for (IIIc) isoforms. Approximately 100% cross-reactivity with recombinant mouse FGF R2, 15% cross-reactivity with recombinant human (rh) FGF RI ( beta isoforms), and no cross-reactivity with rhFGF RI ( alpha isoforms), rhFGF R3, or rhFGF R4 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
98725 | |
Flow Cytometry 10 uL/10^6 cells | |
BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, FGF R2, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2263 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction