Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGFR2 (IIIb) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB665
This item is not returnable.
View return policy
Description
FGFR2 Monoclonal specifically detects FGFR2 in Human samples. It is validated for Western Blot, Neutralization.Specifications
FGFR2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FGFR2 | |
Mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms and S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms | |
500 μg | |
Primary | |
Reacts with multiple isoforms of human FGF R2 although it shows a marked preference for human FGF R2 (IIIb) isoforms in some assays as described below. In direct ELISAs, displays a three-fold preference for FGF R2 (IIIb) isoforms over (IIIc) isoforms. Shows approximately 50% cross-reactivity with recombinant mouse (rm) FGF R2 isoforms and no cross-reactivity with any isoforms of recombinant human (rh) FGF R1 or rhFGF R3. In Western blots, shows approximately 25% cross-reactivity with rhFGF R2 (IIIc) isoforms, 100% cross-reactivity with all isoforms of rmFGF R2 and no cross-reactivity with any isoforms of rhFGF RI, rhFGF R3, or rhFGF R4. Neutralizes the bioactivity of rhFGF R2 (IIIb) isoforms. | |
Human | |
Purified |
Western Blot, Neutralization | |
98707 | |
Western Blot 1 ug/mL, Neutralization 2-8 ug/mL | |
BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, FGF R2, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
2263 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction