Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human FGFR2a (IIIc) Fc Recombinant Protein, PeproTech®

Catalog No. p-7246400
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human FGFR2a (IIIc) is a 65.6 kDa protein containing 589 amino acids. Under reducing conditions, FGFR2a migrates between 100-110 kDa on SDS-PAGE gel. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The FGF family plays a central role during prenatal development and postnatal growth, and the regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. The FGF ligands bind to a family of type I transmembrane tyrosine kinase receptors, which leads to dimerization and activation by sequential autophosphorylation of specific tyrosine residues. Four genes encoding structurally related FGF receptors (FGFR-1 to -4) are known. Alternative splicing of the mRNAs generates numerous forms of FGFR-1 to -3. Alternate forms of FGF receptors can exhibit different specificities with respect to ligand binding. For example, the form designated as FGFR1a (IIc) interacts predominantly with FGF-acidic (FGF1) and FGF-basic (FGF2). A frequent splicing event involving FGFR-1 and -2 results in receptors containing all three Ig domains, referred to as the alpha isoform, or only IgII and IgIII, referred to as thebeta isoform. Only the alpha isoform has been identified for FGFR-3 and FGFR-4. Additional splicing events for FGFR-1 to -3, involving the C-terminal half of the IgIII domain encoded by two mutually exclusive alternative exons, generate FGF receptors with alternative IgIII domains (IIIb and IIIc).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21802
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 2263
Molecular Weight (g/mol) 65.6 kDa
Name Human FGFR2a (IIIc) Fc
Quantity 100 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name FGFR2a (IIIc)
Gene Symbol FGFR2
Biological Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is = 0.1 ng/ml, corresponding to a specific activity of = 1 x 10^7 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence RPSFSLVEDT TLEPEEPPTK YQISQPEVYV AAPGESLEVR CLLKDAAVIS WTKDGVHLGP NNRTVLIGEY LQIKGATPRD SGLYACTASR TVDSETWYFM VNVTDAISSG DDEDDTDGAE DFVSENSNNK RAPYWTNTEK MEKRLHAVPA ANTVKFRCPA GGNPMPTMRW LKNGKEFKQE HRIGGYKVRN QHWSLIMESV VPSDKGNYTC VVENEYGSIN HTYHLDVVER SPHRPILQAG LPANASTVVG GDVEFVCKVY SDAQPHIQWI KHVEKNGSKY GPDGLPYLKV LKAAGVNTTD KEIEVLYIRN VTFEDAGEYT CLAGNSIGIS FHSAWLTVLP APGREKEITA SPDYLEGGPK SCDKTHTCPP CPAPELLGGP SVFLFPPKPK DTLMISRTPE VTCVVVDVSH EDPEVKFNWY VDGVEVHNAK TKPREEQYNS TYRVVSVLTV LHQDWLNGKE YKCKVSNKAL PAPIEKTISK AKGQPREPQV YTLPPSRDEL TKNQVSLTCL VKGFYPSDIA VEWESNGQPE NNYKTTPPVL DSDGSFFLYS KLTVDKSRWQ QGNVFSCSVM HEALHNHYTQ KSLSLSPGK
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.