Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGFR3 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB766G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB766G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB766G100 Supplier R&D Systems Supplier No. FAB766G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGFR3 Monoclonal specifically detects FGFR3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FGFR3
Applications Flow Cytometry
Classification Monoclonal
Clone 136334
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism
Gene Symbols FGFR3
Host Species Mouse
Immunogen Pool of NS0-derived recombinant human FGF R3 alpha (IIIb) and Sf21-derived FGF R3 alpha (IIIc)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2261
Test Specificity Detects the IIIb and IIIc isoforms of human FGF R3 in direct ELISAs and Western blots. Does not cross-react with any isoforms of recombinant mouse (rm) FGF R3, rmFGF R2, recombinant human (rh) FGF R1, rhFGF R2, or rhFGF R4.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.