Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGFR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$212.05 - $2171.79
Specifications
| Antigen | FGFR3 |
|---|---|
| Clone | 136334 |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
Description
FGFR3 Monoclonal specifically detects FGFR3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| FGFR3 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2261 | |
| Mouse myeloma cell line NS0- and Sf21-derived recombinant human FGF R3 alpha (IIIb) | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects the IIIb and IIIc isoforms of human FGF R3 in direct ELISAs and Western blots. Does not cross-react with any isoforms of recombinant mouse (rm) FGF R3, rmFGF R2, recombinant human (rh) FGF R1, rhFGF R2, or rhFGF R4. |
| 136334 | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism | |
| FGFR3 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title