Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGFR3 (IIIb) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1474
This item is not returnable.
View return policy
Description
FGFR3 Monoclonal specifically detects FGFR3 in Human samples. It is validated for Neutralization.Specifications
| FGFR3 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 0.5-2 ug/mL | |
| CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2261 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization | |
| 136315 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGFR3 | |
| Pool of NS0-derived recombinant human FGF R3 alpha (IIIb) and Sf21-derived FGF R3 alpha (IIIc) | |
| 500 μg | |
| Primary | |
| Detects human FGF R3 (IIIb) in direct ELISAs. In direct ELISAs, approximately 25-50% cross-reactivity with recombinant mouse FGF R3 (IIIb) is observed and 12% with recombinant human (rh) FGF R4 is observed and no cross-reactivity with (IIIc) isoforms of rhFGF R3 or any isoforms of rhFGF R1 or rhFGF R2 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction