Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGFR3 (IIIc) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7662100
This item is not returnable.
View return policy
Description
FGFR3 Monoclonal specifically detects FGFR3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| FGFR3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGFR3 | |
| Mouse myeloma cell line NS0-derived recombinant human FGF R3 alpha (IIIc) | |
| 100 μg | |
| Primary | |
| Detects human FGF R3 (IIIc) in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with with recombinant human (rh) FGF R2 (IIIc), recombinant mouse (rm) FGF R2 (IIIc) and rmFGF R3 (IIIc) is observed. In Western blots (non-reducing conditions only), 50-100% cross-reactivity with rhFGF R2 (IIIc), rmFGF R2 (IIIc) and rmFGF R3 (IIIc) is observed. Does not cross-react with the IIIb isoforms of rhFGF R3, rmFGF R3, rhFGF R2 or rmFGF R2. No cross-reactivity with any isoforms of rhFGF RI or rhFGF R4 is observed. | |
| Human | |
| Supernatant |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 136312 | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2261 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction