Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGFR4 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6852R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6852R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6852R100 Supplier R&D Systems Supplier No. FAB6852R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

FGFR4 Monoclonal specifically detects FGFR4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FGFR4
Applications Flow Cytometry
Classification Monoclonal
Clone 240929
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P22455
Gene Alias CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGF R4, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase
Gene Symbols FGFR4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human FGF R4 Leu22-Asp369 (predicted) Accession # P22455
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2264
Test Specificity Detects human FGF R4 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse FGF R4 and recombinant human FGF R5 is observed and no cross-reactivity with any isoform of rhFGF R1, rhFGF R2, or rhFGF R3 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.