Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGFR4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB685SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB685SP 25 μg
MAB685100 100 μg
MAB685500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB685SP Supplier R&D Systems Supplier No. MAB685SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGFR4 Monoclonal specifically detects FGFR4 in Human samples. It is validated for Western Blot.

Specifications

Antigen FGFR4
Applications Western Blot
Classification Monoclonal
Clone 137114
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P22455
Gene Alias CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGF R4, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase
Gene Symbols FGFR4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human FGF R4 Met1-Asp369 Accession # P22455
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2264
Test Specificity Detects human FGF R4 in direct ELISAs and Western blots. In Western blots, this antibody shows no cross-reactivity with any isoform of recombinant human (rh) FGF R1, rhFGF R2, rhFGF R3, recombinant mouse (rm) FGF R2, or rmFGF R3.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.