Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGFR5/FGFRL1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2257
This item is not returnable.
View return policy
Description
FGFR5/FGFRL1 Polyclonal specifically detects FGFR5/FGFRL1 in Human samples. It is validated for Western Blot.Specifications
FGFR5/FGFRL1 | |
Polyclonal | |
Unconjugated | |
Q8N441 | |
FGFRL1 | |
Mouse myeloma cell line NS0-derived recombinant human FGF R5/FGFRL1 Ala25-Pro378 Accession # Q8N441 | |
100 μg | |
Primary | |
Detects human FGF R5/FGFRL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 65% cross-reactivity with recombinant mouse (rm) FGF R5 and rmFGF R5 beta is observed and less than 1% cross-reactivity with recombinant human (rh) FGF R1 alpha (IIIb), rhFGF R1 alpha (IIIc), rhFGF R1 beta (IIIb), rhFGF R1 beta (IIIc), rhFGF R2 alpha (IIIb), rhFGF R2 alpha (IIIc), rhFGF R2 beta (IIIb), rhFGF R2 beta (IIIc), rhFGF R3 alpha (IIIb), rhFGF R3 (IIIc), and rhFGF R4 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
FGF homologous factor receptor, FGF R5, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFRL1, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1 | |
Goat | |
Affinity Purified | |
RUO | |
53834 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction