Learn More
Human FGFR5/FGFRL1 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Supplier:  R&D Systems AF2257SP
 View more versions of this product
Catalog No. AF2257SP
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
FGFR5/FGFRL1 Polyclonal specifically detects FGFR5/FGFRL1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
FGFR5/FGFRL1
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
FGF homologous factor receptor, FGF R5, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFRL1, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1
Goat
Affinity Purified
RUO
53834
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.1 ug/mL
Q8N441
FGFRL1
Mouse myeloma cell line NS0-derived recombinant human FGF R5/FGFRL1 Ala25-Pro378 Accession # Q8N441
25 μg
Primary
Detects human FGF R5/FGFRL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 65% cross-reactivity with recombinant mouse (rm) FGF R5 and rmFGF R5 beta is observed and less than 1% cross-reactivity with recombinant human (rh) FGF R1 alpha (IIIb), rhFGF R1 alpha (IIIc), rhFGF R1 beta (IIIb), rhFGF R1 beta (IIIc), rhFGF R2 alpha (IIIb), rhFGF R2 alpha (IIIc), rhFGF R2 beta (IIIb), rhFGF R2 beta (IIIc), rhFGF R3 alpha (IIIb), rhFGF R3 (IIIc), and rhFGF R4 is observed.
Human
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only