Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGFR5/FGFRL1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2257
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2257 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2257 Supplier R&D Systems Supplier No. BAF2257
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FGFR5/FGFRL1 Polyclonal specifically detects FGFR5/FGFRL1 in Human samples. It is validated for Western Blot.

Specifications

Antigen FGFR5/FGFRL1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q8N441
Gene Alias FGF homologous factor receptor, FGF R5, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFRL1, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1
Gene Symbols FGFRL1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human FGF R5/FGFRL1 Ala25-Pro378 Accession # Q8N441
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53834
Test Specificity Detects human FGF R5/FGFRL1 in Western blots. In Western blots, approximately 60% cross-reactivity with recombinant mouse (rm) FGF R5 and rmFGF R5 beta is observed and less than 1% cross-reactivity with recombinant human (rh) FGF R1 alpha (IIIb), rhFGF R1 alpha (IIIc), rhFGF R1 beta (IIIb), rhFGF R1 beta (IIIc), rhFGF R2 alpha (IIIb), rhFGF R2 alpha (IIIc), rhFGF R2 beta (IIIb), rhFGF R2 beta (IIIc), rhFGF R3 alpha (IIIb), rhFGF R3 (IIIc), and rhFGF R4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.