Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FHL1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5938
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5938 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5938 Supplier R&D Systems Supplier No. AF5938
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FHL1 Polyclonal specifically detects FHL1 in Human samples. It is validated for Western Blot.

Specifications

Antigen FHL1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q13642
Gene Alias bA535K18.1, four and a half LIM domains 1, HLH1, HPLH1, KYOT, KYO-T, LIM protein SLIMMER, skeletal muscle LIM-protein1, SLIM1, SLIM1Isl-1 and Mec-3 domains 1, SLIMMER, XMPMA
Gene Symbols FHL1
Host Species Goat
Immunogen E. coli-derived recombinant human FHL1 Thr93-Asn166 Accession # Q13642
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2273
Test Specificity Detects human FHL1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) FHL2 and rhFHL3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.