Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Fibrillin 1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB301103 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301103 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301103 Supplier Novus Biologicals™ Supplier No. NBP325945
Add to Cart
Add to Cart

Specifications

Assay Range 0.31 to 20 ng/mL
Assay Sensitivity 0.19 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Other Biological Fluids
Specificity This kit recognizes Human Fibrillin 1 in samples. No significant cross-reactivity or interference between Human Fibrillin 1 and analogues was observed
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 2200
Gene Symbol FBN1
Quantity 1 Kit
Research Discipline Extracellular Matrix
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Asprosin, FBN, fibrillin 1, fibrillin 1 (Marfan syndrome), fibrillin 15, fibrillin-1, MASS, MFS1, OCTD, SGS, SSKS, WMS
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.