Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fibrinogen A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9898SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9898SP 25 μg
MAB9898100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9898SP Supplier R&D Systems Supplier No. MAB9898SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fibrinopeptide A Monoclonal specifically detects Fibrinopeptide A in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Fibrinopeptide A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 949330
Conjugate Unconjugated
Dilution Western Blot 1 μg;/mL, Immunohistochemistry 5-25 μg;/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P02671
Gene Alias FGA, Fib2, fibrinogen alpha chain, Fibrinogen alpha subunit, Fibrinogen Type I, fibrinogen, A alpha polypeptide, FPA, MGC119422, MGC119423, MGC119425
Host Species Mouse
Immunogen Synthetic peptide containing human Fibrinogen A Peptide, Accession No. P02671
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 2243.0
Test Specificity Detects human Fibrinogen A in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.