Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fibrinogen Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9404100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9404100 100 μg
MAB9404SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9404100 Supplier R&D Systems Supplier No. MAB9404100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fibrinogen Monoclonal antibody specifically detects Fibrinogen in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Fibrinogen
Applications Immunocytochemistry
Classification Monoclonal
Clone 949332
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P02671
Gene Alias Factor I, Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425
Host Species Mouse
Immunogen Human serum-derived Fibrinogen, Accession # P02671
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2243
Test Specificity Detects human Fibrinogen in direct ELISAs. In Western blots, does not detect Fibrinogen A.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.