Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fibrinogen Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4786
This item is not returnable.
View return policy
Description
Fibrinogen Polyclonal specifically detects Fibrinogen in Human samples. It is validated for Western Blot, Simple Western.Specifications
Fibrinogen | |
Polyclonal | |
Unconjugated | |
Factor I, Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425 | |
Sheep | |
Affinity Purified | |
RUO | |
2243 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL, Simple Western 5 ug/mL | |
FGB | |
Human serum-derived Fibrinogen | |
100 μg | |
Primary | |
Detects human Fibrinogen in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction