Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fibroblast Activation Protein alpha/FAP Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3715U100UG
This item is not returnable.
View return policy
Description
Fibroblast Activation Protein alpha/FAP Monoclonal specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Flow Cytometry.Specifications
Fibroblast Activation Protein alpha/FAP | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q12884 | |
FAP | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Fibroblast Activation Protein alpha/FAP Leu26-Asp760 Accession # Q12884 | |
RUO | |
2191 | |
Human | |
IgG1 |
Flow Cytometry | |
427819 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
Mouse | |
100 μg | |
Primary | |
Detects human Fibroblast Activation Protein alpha/FAP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human DPP6 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction