Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fibroblast Activation Protein alpha/FAP Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3715
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3715 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3715 Supplier R&D Systems Supplier No. BAF3715
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 2 publications

Fibroblast Activation Protein alpha/FAP Polyclonal specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen Fibroblast Activation Protein alpha/FAP
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.5 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. Q12884
Gene Alias 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin
Gene Symbols FAP
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Fibroblast Activation Protein alpha/FAP Leu26-Asp760 Accession # Q12884
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2191
Test Specificity Detects human Fibroblast Activation Protein alpha/FAP in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human DPPIV is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.