Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fibromodulin/FMOD Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5945
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5945 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5945 Supplier R&D Systems Supplier No. MAB5945
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fibromodulin/FMOD Monoclonal specifically detects Fibromodulin/FMOD in Human samples. It is validated for Western Blot.

Specifications

Antigen Fibromodulin/FMOD
Applications Western Blot
Classification Monoclonal
Clone 549302
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_002014
Gene Alias Collagen-binding 59 kDa protein, fibromodulin, fibromodulin proteoglycan, FM, FMOD, Keratan sulfate proteoglycan fibromodulin, SLRR2E, SLRR2EKSPG fibromodulin
Gene Symbols FMOD
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fibromodulin/FMOD Asp75-Ile376 Accession # NP_002014
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2331
Test Specificity Detects human Fibromodulin/FMOD in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Lumican is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.