Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Fibronectin ELISA Kit (Colorimetric)

Click to view available options
Quantity:
96 Tests
Description
The Human Fibronectin ELISA Kit (Colorimetric) measures human Fibronectin in cell culture supernatant, serum, plasma (edta, heparin), urine.
Specifications
Specifications
Assay Range | 0.31 - 20.0ng/mL |
Assay Sensitivity | 0.1ng/mL |
Product Type | ELISA Kit |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine |
Specificity | The interference of circulating factors of the immune systemwas evaluated by spiking these proteins at physiologically relevant concentrations into positive serum. There was no crossreactivity detected. |
Target Species | Human |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 2335 |
Gene Symbol | FN1 |
Kit Contents | Biotin-Conjugate anti-human Fibronectin polyclonal antibody, Streptavidin-HRP, Human Fibronectin Standard lyophilized, 40ng/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films, Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to human Fibronectin |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction