Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fibulin 3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6235
This item is not returnable.
View return policy
Description
Fibulin-3/EFEMP1 Polyclonal specifically detects Fibulin-3/EFEMP1 in Human samples. It is validated for Western Blot.Specifications
Fibulin-3/EFEMP1 | |
Polyclonal | |
Unconjugated | |
Q12805 | |
EFEMP1 | |
Chinese hamster ovary cell line CHO-derived recombinant human Fibulin 3 Gln19-Phe493 Accession # Q12805 | |
100 μg | |
Primary | |
Detects human Fibulin 3 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
DHRD, EFEMP1, EGF containing fibulin-like extracellular matrix protein 1, EGF-containing fibulin-like extracellular matrix protein 1, Extracellular protein S1-5, FBLN3, FBLN3DRAD, FBNLFLJ35535, FIBL-3, fibrillin-like, Fibrillin-like protein, Fibulin-3, MGC111353, MLVT, MTLV, S1-5 | |
Sheep | |
Affinity Purified | |
RUO | |
2202 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction