Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fibulin 5/DANCE Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB3095SP
This item is not returnable.
View return policy
Description
Fibulin 5/DANCE Monoclonal specifically detects Fibulin 5/DANCE in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Fibulin 5/DANCE | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q9UBX5 | |
FBLN5 | |
Mouse myeloma cell line NS0-derived recombinant human Fibulin 5/DANCE Gln24-Phe448 Accession # Q9UBX5 | |
25 μg | |
Primary | |
Detects human Fibulin 5/DANCE in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
293904 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ARMD3fibulin-5, Dance, DANCEFIBL-5, Developmental arteries and neural crest EGF-like protein, EVEC, FBLN5, fibulin 5, UP50, UP50FLJ90059, Urine p50 protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10516 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction