Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ficolin-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2428SP
This item is not returnable.
View return policy
Description
Ficolin-2 Polyclonal specifically detects Ficolin-2 in Human samples. It is validated for Western Blot.Specifications
Ficolin-2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
37 kDa elastin-binding protein, Collagen/fibrinogen domain-containing protein 2, EBP-37ficolin (collagen/fibrinogen domain-containing lectin) 2 (hucolin), FCN2, FCNL, FCNLficolin B, ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin), ficolin-2, Ficolin2, Ficolin-B, Ficolin-beta, Hucolin, L-Ficolin, P35, Serum lectin p35 | |
Goat | |
Affinity Purified | |
RUO | |
2220 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q15485 | |
FCN2 | |
Mouse myeloma cell line NS0-derived recombinant human Ficolin-2 Leu26-Ala313 Accession # Q15485 | |
25 μg | |
Primary | |
Detects human Ficolin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human Ficolin-3 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction