Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FKBP12 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $526.00
Specifications
Antigen | FKBP12 |
---|---|
Clone | 1049713 |
Dilution | Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB37771SP
![]() |
R&D Systems
MAB37771SP |
25 μg |
Each for $200.50
|
|
|||||
MAB37771100
![]() |
R&D Systems
MAB37771100 |
100 μg |
Each for $526.00
|
|
|||||
Description
FKBP12 Monoclonal antibody specifically detects FKBP12 in Human samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
FKBP12 | |
Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP1a, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PKC12, PPIASE, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
E. coli-derived human FKBP12F36V protein, Gly1-Glu108, Accession # P62942 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1049713 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P62942 | |
2280 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title