Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FLI1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6474
This item is not returnable.
View return policy
Description
FLI1 Polyclonal specifically detects FLI1 in Human samples. It is validated for Western Blot.Specifications
FLI1 | |
Polyclonal | |
Unconjugated | |
Q01543 | |
FLI1 | |
E. coli-derived recombinant human FLI1 Ser13-Pro112 Accession # Q01543 | |
100 μg | |
Primary | |
Detects human FLI1 in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
ERGB, EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB | |
Sheep | |
Affinity Purified | |
RUO | |
2313 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction