Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FLRT3 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2795
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2795 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2795 Supplier R&D Systems Supplier No. BAF2795
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FLRT3 Polyclonal specifically detects FLRT3 in Human samples. It is validated for Western Blot.

Specifications

Antigen FLRT3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9NZU0
Gene Alias fibronectin leucine rich transmembrane protein 3, Fibronectin-like domain-containing leucine-rich transmembrane protein 3, HH21, KIAA1469, leucine-rich repeat transmembrane protein FLRT3
Gene Symbols FLRT3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human FLRT3 Lys29-Pro528 Accession # Q9NZU0
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23767
Test Specificity Detects human FLRT3 in Western blots. In Western blots, less than 10% cross-reactivity with recombinant human (rh) FLRT1 and rhFLRT2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.